Assay Details
Target Gene Details
Entrez Gene ID: | 73133 |
Gene Name: | RIKEN cDNA 3110021N24 gene |
Gene Aliases: |
2610002D03Rik |
Location: |
Chr.4:108719649-108781904 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 3110021N24Rik | NM_001254730.1 | NP_001241659.1 | ||
| BE649473.1 |
Target Gene Details
Entrez Gene ID: | 230597 |
Gene Name: | zinc finger, FYVE domain containing 9 |
Gene Aliases: |
E030027L17, Madhip, NSP, SARA, SMADIP |
Location: |
Chr.4:108637834-108781148 on Build GRCm38 |
Assay Gene Location: | Overlaps - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfyve9 | NM_183300.2 | NP_899123.2 | ||
| DQ303413.1 |

Genomic Map