Assay Details
Target Gene Details
Entrez Gene ID: | 192656 |
Gene Name: | receptor (TNFRSF)-interacting serine-threonine kinase 2 |
Gene Aliases: |
2210420D18Rik, CARD3, CARDIAK, CCK, D4Bwg0615e, RICK, RIP2 |
Location: |
Chr.4:16122741-16163679 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ripk2 | NM_138952.3 | 4 | 664 | NP_620402.1 |
| AF461040.1 | 4 | 562 | AAL96436.1 | |
| AF487539.1 | 4 | 562 | AAM76919.1 | |
| AK159593.1 | 4 | 688 | BAE35213.1 | |
| AK172319.1 | 4 | 686 | BAE42942.1 | |
| AK172417.1 | 4 | 686 | BAE42997.1 | |
| BC025611.1 | 2 | 182 | ||
| BC069878.1 | 4 | 689 | AAH69878.1 |
More Information
Set Membership: |
|

Genomic Map