Assay Details
Target Gene Details
Entrez Gene ID: | 22062 |
Gene Name: | transformation related protein 73 |
Gene Aliases: |
TAp73, Tp73, deltaNp73, p73 |
Location: |
Chr.4:154056249-154140208 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trp73 | NM_001126330.1 | 8 | 1234 | NP_001119802.1 |
| NM_001126331.1 | 8 | 1234 | NP_001119803.1 | |
| NM_011642.3 | 10 | 1524 | NP_035772.2 | |
| AB221594.1 | 9 | 1116 | ||
| AK014503.1 | 3 | 213 | BAB29401.1 | |
| AK017412.1 | 7 | 765 | BAB30732.1 | |
| AK139633.1 | 8 | 1235 | BAE24089.1 | |
| BC066045.1 | 8 | 1121 | AAH66045.1 | |
| Y19234.1 | 10 | 1525 | CAB81953.1 | |
| Y19235.1 | 8 | 1228 | CAB81954.1 |
More Information
Set Membership: |
|

Genomic Map