Assay Details
Target Gene Details
Entrez Gene ID: | 22275 |
Gene Name: | uroporphyrinogen decarboxylase |
Gene Aliases: |
AI323803, Uro-d |
Location: |
Chr.4:116990212-116994436 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Urod | NM_009478.3 | NP_033504.2 | ||
| AK002844.1 | ||||
| AK007470.1 | ||||
| AK135090.1 | ||||
| AK172426.1 | BAE43002.1 | |||
| BC008109.1 | AAH08109.1 |
More Information
Set Membership: |
|

Genomic Map