Assay Details
Target Gene Details
Entrez Gene ID: | 22062 |
Gene Name: | transformation related protein 73 |
Gene Aliases: |
TAp73, Tp73, deltaNp73, p73 |
Location: |
Chr.4:154056249-154140208 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trp73 | NM_011642.3 | 3 | 518 | NP_035772.2 |
| AB221594.1 | 2 | 110 | ||
| BB663805.1 | 3 | 254 | ||
| Y19234.1 | 3 | 519 | CAB81953.1 |
More Information
Set Membership: |
|

Genomic Map