Assay Details
Target Gene Details
Entrez Gene ID: | 269589 |
Gene Name: | synaptotagmin-like 1 |
Gene Aliases: |
9030417L18, PSGL-1, Slp1 |
Location: |
Chr.4:133253090-133263087 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sytl1 | NM_031393.2 | NP_113570.2 | ||
| AB050741.1 | BAB32651.1 | |||
| AK078868.1 | ||||
| AK131162.1 | BAD21412.1 | |||
| AK154176.1 | ||||
| BC005623.1 | AAH05623.1 |

Genomic Map