Assay Details
Target Gene Details
Entrez Gene ID: | 100504717 |
Gene Name: | predicted gene 12709 |
Gene Aliases: |
OTTMUSG00000008056 |
Location: |
Chr.4:102967266-102989755 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm12709 | NR_040444.1 | 4 | 887 | |
| AK038336.1 | 4 | 888 |
Target Gene Details
Entrez Gene ID: | 73094 |
Gene Name: | SH3-domain GRB2-like (endophilin) interacting protein 1 |
Gene Aliases: |
3110007P09Rik, SGIP1alpha, gag |
Location: |
Chr.4:102759167-102972701 on Build GRCm38 |
Assay Gene Location: | Within Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sgip1 | NM_001285852.1 | 25 | 2919 | NP_001272781.1 |
| NM_001285859.1 | 23 | 2334 | NP_001272788.1 | |
| NM_001285860.1 | 22 | 2274 | NP_001272789.1 | |
| NM_001285862.1 | 19 | 2181 | NP_001272791.1 | |
| NM_144906.2 | 23 | 2775 | NP_659155.1 | |
| AB262964.1 | 24 | 2498 | ||
| AK032439.1 | 22 | 2065 | BAC27870.1 | |
| AK043018.1 | 23 | 2160 | BAC31435.1 | |
| AK148043.1 | 7 | 862 | BAE28309.1 | |
| AK148302.1 | 19 | 1962 | BAE28466.1 | |
| BC017596.1 | 23 | 2547 | AAH17596.1 |
More Information
Set Membership: |
|

Genomic Map