Assay Details
Target Gene Details
Entrez Gene ID: | 68970 |
Gene Name: | DDB1 and CUL4 associated factor 12 |
Gene Aliases: |
1500001L20Rik, 5830424K06Rik, AA420338, AI851081, Wdr40a |
Location: |
Chr.4:41291300-41316726 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dcaf12 | NM_026893.3 | NP_081169.2 | ||
| AK005099.1 | BAB23817.2 | |||
| AK030173.1 | BAC26822.1 | |||
| AK031089.1 | BAC27247.1 | |||
| AK078036.1 | ||||
| AK087517.1 | BAC39908.1 | |||
| AK090028.1 | BAC41053.1 | |||
| AK143635.1 | ||||
| AK173289.1 | BAD32567.1 | |||
| BC023852.1 | AAH23852.1 |

Genomic Map