Assay Details
Target Gene Details
| Entrez Gene ID: | 66096 | 
| Gene Name: | late endosomal/lysosomal adaptor, MAPK and MTOR activator 4 | 
| Gene Aliases: | 0910001L09Rik, AV006840 | 
| Location: | Chr.5:138255482-138259395 on Build GRCm38 | 
| Assay Gene Location: | Overlaps Intron 2 - Exon 3 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| Lamtor4 | NM_001081108.2 | NP_001074577.1 | ||
| AK002488.1 | BAC24993.1 | |||
| AK003086.1 | BAC25020.1 | |||
| AK003250.1 | ||||
| AK010598.1 | BAC25306.1 | |||
| AK146994.1 | ||||
| BC144958.1 | 
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top Intragenic
							Intragenic