Assay Details
Target Gene Details
Entrez Gene ID: | 73212 |
Gene Name: | RIKEN cDNA 3110082I17 gene |
Gene Aliases: |
AW411904 |
Location: |
Chr.5:139359739-139460534 on Build GRCm38 |
Assay Gene Location: | Overlaps - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 3110082I17Rik | NM_028469.3 | NP_082745.2 | ||
| AK014271.1 | BAB29235.1 | |||
| AV470085.1 | ||||
| BU562762.1 |

Genomic Map