Assay Details
Target Gene Details
Entrez Gene ID: | 21936 |
Gene Name: | tumor necrosis factor receptor superfamily, member 18 |
Gene Aliases: |
AITR, Gitr |
Location: |
Chr.4:156026342-156028891 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tnfrsf18 | NM_009400.2 | NP_033426.1 | ||
| NM_021985.2 | NP_068820.1 | |||
| AF229432.1 | AAF61566.1 | |||
| AF229433.1 | AAF61567.1 | |||
| AF229434.1 | AAF61568.1 | |||
| AK020762.1 | BAC25639.1 | |||
| AK081878.1 | BAC38357.1 | |||
| AY157832.1 | AAO21307.1 | |||
| AY157833.1 | AAO21308.1 | |||
| U82534.1 | AAB81243.1 |
Target Gene Details
Entrez Gene ID: | 330010 |
Gene Name: | tubulin tyrosine ligase-like family, member 10 |
Gene Aliases: |
4833412E22Rik, 4930595O22Rik, AU022799, BC042775, Ttll5 |
Location: |
Chr.4:156023748-156084440 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ttll10 |
More Information
Set Membership: |
|

Genomic Map