Assay Details
Target Gene Details
Entrez Gene ID: | 22163 |
Gene Name: | tumor necrosis factor receptor superfamily, member 4 |
Gene Aliases: |
ACT35, CD134, Ly-70, Ox40, TXGP1L, Txgp1 |
Location: |
Chr.4:156013695-156016613 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tnfrsf4 | NM_011659.2 | NP_035789.1 | ||
| BC139239.1 | ||||
| BC139266.1 | ||||
| Z21674.1 | CAA79772.1 |
More Information
Set Membership: |
|

Genomic Map