Assay Details
Target Gene Details
Entrez Gene ID: | 77593 |
Gene Name: | ubiquitin specific petidase 45 |
Gene Aliases: |
3110003C05Rik, 4930550B20Rik, AI843191, Gcap7 |
Location: |
Chr.4:21767161-21837872 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Usp45 | NM_001290425.1 | NP_001277354.1 | ||
| NM_152825.2 | NP_690038.1 | |||
| AK019756.1 | ||||
| AK040660.1 | ||||
| AK088073.1 | BAC40129.1 | |||
| AK162825.1 | ||||
| BB213271.2 | ||||
| BC027768.1 | AAH27768.1 |
More Information
Set Membership: |
|

Genomic Map