Assay Details
Target Gene Details
Entrez Gene ID: | 75369 |
Gene Name: | RIKEN cDNA 4930563F08 gene |
Gene Aliases: |
- |
Location: |
Chr.5:131880269-131883768 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4930563F08Rik | NR_040704.1 | |||
| AK016203.1 |
Target Gene Details
Entrez Gene ID: | 319974 |
Gene Name: | autism susceptibility candidate 2 |
Gene Aliases: |
2700063G02Rik, A730011F23Rik, D830032G16Rik |
Location: |
Chr.5:131437682-132542343 on Build GRCm38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Auts2 | NM_177047.3 | NP_796021.2 | ||
| AK052418.1 | BAC34980.1 | |||
| BC151031.1 | ||||
| BC151047.1 |
More Information
Set Membership: |
|

Genomic Map