Assay Details
Target Gene Details
Entrez Gene ID: | 320129 |
Gene Name: | adrenergic receptor kinase, beta 2 |
Gene Aliases: |
4833444A01Rik, AI851927, AW551196, Adrbk-2, Bark-2, GRK3 |
Location: |
Chr.5:112910478-113015951 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adrbk2 | NM_001285806.1 | NP_001272735.1 | ||
| NM_177078.4 | NP_796052.2 | |||
| AK076533.1 | BAC36382.1 | |||
| AK134672.1 | BAE22235.1 | |||
| AK141186.1 |
More Information
Set Membership: |
|

Genomic Map