Assay Details
Target Gene Details
Entrez Gene ID: | 269700 |
Gene Name: | predicted gene 15800 |
Gene Aliases: |
A130010L24, AU042671, BC019639, EG545802, OTTMUSG00000026283, mKIAA0614 |
Location: |
Chr.5:121219339-121368577 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm15800 | NM_181421.4 | NP_852086.3 |
More Information
Set Membership: |
|

Genomic Map