Assay Details
Target Gene Details
Entrez Gene ID: | 231600 |
Gene Name: | checkpoint with forkhead and ring finger domains |
Gene Aliases: |
5730484M20Rik, C230082M18, RNF116 |
Location: |
Chr.5:110135759-110171973 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 17 - Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Chfr | NM_001289577.1 | NP_001276506.1 | ||
| NM_001289578.1 | NP_001276507.1 | |||
| NM_001289579.1 | NP_001276508.1 | |||
| NM_001289580.1 | NP_001276509.1 | |||
| NM_172717.4 | NP_766305.2 | |||
| AK048930.1 | ||||
| AK077629.1 | BAC36912.1 | |||
| AK147892.1 | BAE28209.1 | |||
| AK148436.1 | ||||
| AK154038.1 | BAE32331.1 | |||
| AK155295.1 | BAE33172.1 | |||
| AK155525.1 | BAE33309.1 | |||
| AK169182.1 | BAE40960.1 | |||
| AK169193.1 | BAE40969.1 | |||
| BC049792.1 | AAH49792.1 |
Target Gene Details
Entrez Gene ID: | 100504007 |
Gene Name: | predicted gene 15787 |
Gene Aliases: |
OTTMUSG00000026209 |
Location: |
Chr.5:110167508-110176504 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm15787 | NR_040430.1 | 3 | 679 | |
| NR_040431.1 | 3 | 619 | ||
| AK053084.1 | 3 | 620 |
More Information
Set Membership: |
|

Genomic Map