Assay Details
Target Gene Details
Entrez Gene ID: | 57423 |
Gene Name: | ATP synthase, H+ transporting, mitochondrial F0 complex, subunit F2 |
Gene Aliases: |
1110019H14Rik |
Location: |
Chr.5:145183706-145191592 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atp5j2 | NM_020582.2 | NP_065607.1 | ||
| AB030192.1 | BAA92756.1 | |||
| AK002519.1 | BAB22157.1 | |||
| AK003235.1 | BAB22660.1 | |||
| AK003817.1 | BAB23014.1 | |||
| AK013130.1 | BAB28667.1 | |||
| AK151737.1 | BAE30651.1 | |||
| AK168095.1 | BAE40067.1 | |||
| BC029226.1 | AAH29226.1 |
More Information
Set Membership: |
|

Genomic Map