Assay Details
Target Gene Details
Entrez Gene ID: | 20309 |
Gene Name: | chemokine (C-X-C motif) ligand 15 |
Gene Aliases: |
Il8, Scyb15, lungkine, weche |
Location: |
Chr.5:90794534-90803067 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cxcl15 | NM_011339.2 | NP_035469.1 | ||
| AF082859.1 | AAD38079.1 | |||
| AK142955.1 | BAE25229.1 | |||
| BC061138.1 | AAH61138.1 |
More Information
Set Membership: |
|

Genomic Map