Assay Details
Target Gene Details
Entrez Gene ID: | 73212 |
Gene Name: | RIKEN cDNA 3110082I17 gene |
Gene Aliases: |
AW411904 |
Location: |
Chr.5:139359739-139460534 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 3110082I17Rik | NM_028469.3 | NP_082745.2 | ||
| AK014271.1 | BAB29235.1 | |||
| AV470085.1 | ||||
| BU562762.1 |
Target Gene Details
Entrez Gene ID: | 319772 |
Gene Name: | RIKEN cDNA C130050O18 gene |
Gene Aliases: |
- |
Location: |
Chr.5:139403597-139416092 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C130050O18Rik | NM_177000.3 | 1 | 97 | NP_795974.2 |
| AK048344.1 | 1 | 46 | BAC33306.1 | |
| AK086625.1 | 1 | 36 | ||
| AK155269.1 | 1 | 68 | BAE33154.1 | |
| AK170398.1 | ||||
| AK171460.1 | 1 | 98 | ||
| BC100320.1 | 2 | 180 | AAI00321.1 | |
| BY199357.1 | 1 | 99 |
More Information
Set Membership: |
|

Genomic Map