Assay Details
Target Gene Details
Entrez Gene ID: | 545817 |
Gene Name: | cytochrome P450, family 2, subfamily w, polypeptide 1 |
Gene Aliases: |
Gm455 |
Location: |
Chr.5:139352617-139359607 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyp2w1 | NM_001160265.1 | 6 | 921 | NP_001153737.1 |
More Information
Set Membership: |
|

Genomic Map