Assay Details
Target Gene Details
Entrez Gene ID: | 231841 |
Gene Name: | BRCA1-associated ATM activator 1 |
Gene Aliases: |
AA881470, Baat1 |
Location: |
Chr.5:140704786-140719379 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Brat1 | NM_001276287.1 | NP_001263216.1 | ||
| NM_181066.3 | NP_851411.1 | |||
| NR_074085.1 | ||||
| AF226663.1 | AAF43013.1 | |||
| AK049321.1 | ||||
| AK084987.1 | BAC39330.1 | |||
| AK085091.1 | BAC39362.1 | |||
| AK154448.1 | BAE32593.1 | |||
| AK170269.1 | BAE41674.1 | |||
| BC068301.1 | AAH68301.1 |

Genomic Map