Assay Details
Target Gene Details
Entrez Gene ID: | 80752 |
Gene Name: | family with sequence similarity 20, member C |
Gene Aliases: |
C76981, DMP-4, DMP4, GEF-CK, mKIAA4081 |
Location: |
Chr.5:138754306-138810072 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam20c | NM_030565.6 | NP_085042.2 | ||
| AK149780.1 | ||||
| AK154810.1 | ||||
| AK220286.1 | BAD90211.1 | |||
| AY778962.1 | AAV91326.1 | |||
| BC004044.1 | ||||
| BC025814.1 | AAH25814.1 | |||
| BC025826.1 | AAH25826.1 | |||
| CT010363.1 | CAJ18570.1 |

Genomic Map