Assay Details
Target Gene Details
Entrez Gene ID: | 433868 |
Gene Name: | RIKEN cDNA 3110082J24 gene |
Gene Aliases: |
ENSMUSG00000073106 |
Location: |
Chr.5:30103584-30106086 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 3110082J24Rik | NM_001256263.1 | NP_001243192.1 | ||
| AK132123.1 | BAE20991.1 |
Target Gene Details
Entrez Gene ID: | 242915 |
Gene Name: | GRB2 associated, regulator of MAPK1-like |
Gene Aliases: |
Fam59b, Gm444 |
Location: |
Chr.5:30105196-30118380 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gareml | NM_001167879.1 | NP_001161351.1 | ||
| CX204487.1 | ||||
| CX237000.1 |

Genomic Map