Assay Details
Target Gene Details
Entrez Gene ID: | 94244 |
Gene Name: | FK506 binding protein 6 |
Gene Aliases: |
1700008G22Rik, 36kDa, AU017274, D5Ertd724e, FKBP-36, FKBP-6 |
Location: |
Chr.5:135291704-135351807 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fkbp6 | NM_001277891.1 | 4 | 385 | NP_001264820.1 |
| NM_001277892.1 | 4 | 332 | NP_001264821.1 | |
| NM_001277893.1 | 4 | 385 | NP_001264822.1 | |
| NM_033571.3 | 4 | 385 | NP_291049.1 | |
| AF367709.1 | 4 | 366 | AAK53411.1 | |
| AF367710.1 | 4 | 332 | AAK53412.1 | |
| AF367711.1 | 4 | 332 | AAK53413.1 | |
| AK090036.1 | 4 | 376 | BAC41058.1 | |
| AY029192.1 | 4 | 332 | AAK39645.1 | |
| BC139092.1 | 4 | 385 | ||
| BC139097.1 | 4 | 385 |
More Information
Set Membership: |
|

Genomic Map