Assay Details
Target Gene Details
Entrez Gene ID: | 68017 |
Gene Name: | FtsJ homolog 2 (E. coli) |
Gene Aliases: |
2310037B18Rik |
Location: |
Chr.5:140327674-140331898 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ftsj2 | NM_026510.1 | NP_080786.1 | ||
| AK009656.1 | BAB26420.1 | |||
| AK017300.1 | BAB30680.1 | |||
| AK077929.1 | BAC37070.1 | |||
| AK145808.1 | BAE26662.1 |
Target Gene Details
Entrez Gene ID: | 17766 |
Gene Name: | nudix (nucleoside diphosphate linked moiety X)-type motif 1 |
Gene Aliases: |
Mth1 |
Location: |
Chr.5:140321627-140338137 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nudt1 |
More Information
Set Membership: |
|

Genomic Map