Assay Details
Target Gene Details
Entrez Gene ID: | 231659 |
Gene Name: | GCN1 general control of amino-acid synthesis 1-like 1 (yeast) |
Gene Aliases: |
4932409G22, AL022764, G431004K08Rik, GCN1L, mKIAA0219 |
Location: |
Chr.5:115565219-115622654 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gcn1l1 | NM_172719.2 | NP_766307.2 | ||
| AK037425.1 | ||||
| AK041717.1 | ||||
| AK090137.1 | ||||
| AK147257.1 | BAE27802.1 | |||
| AK154512.1 | BAE32641.1 | |||
| AK158040.1 | ||||
| BC082548.1 | AAH82548.1 | |||
| BC150735.1 | ||||
| CD350892.1 |
More Information
Set Membership: |
|

Genomic Map