Assay Details
Target Gene Details
Entrez Gene ID: | 22791 |
Gene Name: | DnaJ (Hsp40) homolog, subfamily C, member 2 |
Gene Aliases: |
AU020218, MIDA1, Zrf1, Zrf2 |
Location: |
Chr.5:21757264-21785248 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dnajc2 | NM_009584.4 | NP_033610.1 | ||
| AK013295.1 | ||||
| AK131964.1 | BAE20907.1 | |||
| AK136951.1 | BAE23185.1 | |||
| AK161347.1 | BAE36340.1 | |||
| AK161376.1 | BAE36356.1 | |||
| AK162409.1 | BAE36900.1 | |||
| AK167991.1 | ||||
| AK169850.1 | BAE41409.1 | |||
| AK215516.1 | ||||
| BC052027.1 | AAH52027.1 | |||
| D63784.1 | BAA09854.1 | |||
| U53208.1 | AAC52486.1 |
More Information
Set Membership: |
|

Genomic Map