Assay Details
Target Gene Details
Entrez Gene ID: | 231098 |
Gene Name: | DnaJ (Hsp40) homolog, subfamily C, member 5 gamma |
Gene Aliases: |
4922503E23Rik |
Location: |
Chr.5:31108292-31112526 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dnajc5g | NM_177677.3 | NP_808345.1 | ||
| AK076638.1 | BAC36428.1 | |||
| BC089502.1 | AAH89502.1 |
More Information
Set Membership: |
|

Genomic Map