Assay Details
Target Gene Details
Entrez Gene ID: | 15234 |
Gene Name: | hepatocyte growth factor |
Gene Aliases: |
C230052L06Rik, HGF/SF, NK1, NK2, SF, SF/HGF |
Location: |
Chr.5:16553335-16622408 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hgf | NM_001289458.1 | 9 | 1160 | NP_001276387.1 |
| NM_001289459.1 | 9 | 1149 | NP_001276388.1 | |
| NM_001289461.1 | 8 | 967 | NP_001276390.1 | |
| NM_010427.5 | 9 | 1132 | NP_034557.3 | |
| AK042121.1 | 9 | 1078 | BAC31175.1 | |
| BC119228.1 | 9 | 1019 | ||
| D10212.1 | 8 | 955 | BAA01064.1 | |
| D10213.1 | 8 | 940 | BAA01065.1 | |
| S71816.1 | 8 | 955 | AAB31855.1 | |
| X72307.1 | 8 | 1016 | CAA51054.1 | |
| X84046.1 | 8 | 995 | CAA58865.1 |
More Information
Set Membership: |
|

Genomic Map