Assay Details
Target Gene Details
Entrez Gene ID: | 54426 |
Gene Name: | hepatocyte growth factor activator |
Gene Aliases: |
HGFA |
Location: |
Chr.5:35041514-35048436 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hgfac | NM_019447.2 | 2 | 203 | NP_062320.2 |
| AB009294.1 | 2 | 203 | ||
| AF099017.1 | 2 | 163 | AAF02489.1 | |
| AF224724.1 | 2 | 163 | AAF34712.1 | |
| AK007987.1 | 2 | 201 | BAB25391.1 | |
| AK075718.1 | 2 | 111 | ||
| AK144554.1 | 2 | 175 | ||
| AK149522.1 | 2 | 167 | BAE28936.1 | |
| AK165433.1 | 2 | 180 | ||
| AK169989.1 | 2 | 199 | BAE41500.1 | |
| BC019376.1 | 2 | 168 | AAH19376.1 | |
| BY202513.1 | 2 | 246 |
More Information
Set Membership: |
|

Genomic Map