Assay Details
Target Gene Details
Entrez Gene ID: | 70592 |
Gene Name: | RIKEN cDNA 5730480H06 gene |
Gene Aliases: |
- |
Location: |
Chr.5:48395573-48414294 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 5730480H06Rik | NR_045500.1 | |||
| AK017706.1 |
Target Gene Details
Entrez Gene ID: | 80334 |
Gene Name: | Kv channel interacting protein 4 |
Gene Aliases: |
AV032399, Calp, Calp250, KChIP4, KChIP4a |
Location: |
Chr.5:48389503-49525122 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kcnip4 | NM_001199242.1 | NP_001186171.1 | ||
| NM_001199243.1 | NP_001186172.1 | |||
| NM_001199244.1 | NP_001186173.1 | |||
| NM_001199245.1 | NP_001186174.1 | |||
| NM_030265.3 | NP_084541.3 | |||
| AF305071.1 | AAG36976.1 | |||
| AF453243.1 | AAL86766.1 | |||
| AK039048.1 | BAC30218.1 | |||
| AK049662.1 | ||||
| AK133416.1 | BAE21645.1 | |||
| AK148685.1 | BAE28641.1 | |||
| AK165773.1 | BAE38374.1 | |||
| AY647240.1 | AAT68466.1 | |||
| BC051130.1 | AAH51130.1 | |||
| DQ148501.1 | AAZ77818.1 | |||
| DQ148502.1 | AAZ77819.1 | |||
| DQ148503.1 | AAZ77820.1 | |||
| DQ148504.1 | AAZ77821.1 |
More Information
Set Membership: |
|

Genomic Map