Assay Details
Target Gene Details
Entrez Gene ID: | 74254 |
Gene Name: | GPN-loop GTPase 1 |
Gene Aliases: |
2410004J02Rik, AI449615, MBDIN, NTPBP, Xab1 |
Location: |
Chr.5:31494752-31511631 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gpn1 | NM_133756.4 | 7 | 488 | NP_598517.1 |
| AK010393.1 | 7 | 502 | ||
| AK077645.1 | 7 | 489 | BAC36923.1 | |
| AK163635.1 | 5 | 2817 | ||
| BC020174.1 | 7 | 488 | AAH20174.1 | |
| BC096466.1 | 7 | 481 | AAH96466.1 |
More Information
Set Membership: |
|

Genomic Map