Assay Details
Target Gene Details
Entrez Gene ID: | 66437 |
Gene Name: | fission 1 (mitochondrial outer membrane) homolog (yeast) |
Gene Aliases: |
2010003O14Rik, Ttc11 |
Location: |
Chr.5:136953264-136966234 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fis1 | NM_001163243.1 | NP_001156715.1 | ||
| NM_025562.3 | NP_079838.1 | |||
| AK004479.1 | BAB23324.1 | |||
| AK004917.1 | BAB23668.1 | |||
| AK008078.1 | BAB25445.1 | |||
| AK008323.1 | BAB25601.1 | |||
| AK008916.1 | BAB25966.1 | |||
| AW988976.1 | ||||
| BC010783.1 | AAH10783.1 | |||
| CA322565.1 | ||||
| CD774298.1 |

Genomic Map