Assay Details
Target Gene Details
Entrez Gene ID: | 110391 |
Gene Name: | quinoid dihydropteridine reductase |
Gene Aliases: |
2610008L04Rik, D5Ertd371e, Dhpr, PKU2 |
Location: |
Chr.5:45434032-45450229 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Qdpr | NM_024236.2 | NP_077198.1 | ||
| AK011351.1 | BAB27559.1 | |||
| AK078123.1 | BAC37135.1 | |||
| AK159107.1 | BAE34823.1 | |||
| AK161660.1 | BAE36516.1 | |||
| AK166694.1 | BAE38951.1 | |||
| AK214053.1 | ||||
| BC002107.1 | AAH02107.1 |
More Information
Set Membership: |
|

Genomic Map