Assay Details
Target Gene Details
Entrez Gene ID: | 56744 |
Gene Name: | platelet factor 4 |
Gene Aliases: |
Cxcl4, Scyb4 |
Location: |
Chr.5:90772435-90773383 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pf4 | NM_019932.4 | NP_064316.1 | ||
| AB017491.2 | BAA75660.1 | |||
| AK159278.1 | BAE34956.1 | |||
| AK160040.1 | BAE35582.1 | |||
| BC061111.1 | AAH61111.1 | |||
| BY794492.1 |
More Information
Set Membership: |
|

Genomic Map