Assay Details
Target Gene Details
Entrez Gene ID: | 56744 |
Gene Name: | platelet factor 4 |
Gene Aliases: |
Cxcl4, Scyb4 |
Location: |
Chr.5:90772435-90773383 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pf4 | NM_019932.4 | 3 | 443 | NP_064316.1 |
| AB017491.2 | 3 | 378 | BAA75660.1 | |
| AK159278.1 | 3 | 443 | BAE34956.1 | |
| AK160040.1 | 3 | 443 | BAE35582.1 | |
| BC061111.1 | 3 | 375 | AAH61111.1 |
More Information
Set Membership: |
|

Genomic Map