Assay Details
Target Gene Details
Entrez Gene ID: | 53324 |
Gene Name: | neuronal pentraxin 2 |
Gene Aliases: |
Narp, np2 |
Location: |
Chr.5:144545887-144557478 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nptx2 | NM_016789.3 | 4 | 1102 | NP_058069.1 |
| AF049124.1 | 4 | 992 | AAC05131.1 | |
| AK134806.1 | 4 | 1103 | BAE22293.1 | |
| BC026054.1 | 4 | 1097 | AAH26054.1 | |
| DV649643.1 | 4 | 541 |

Genomic Map