Assay Details
Target Gene Details
Entrez Gene ID: | 107939 |
Gene Name: | nuclear pore membrane protein 121 |
Gene Aliases: |
2610027A18Rik, C80273, mKIAA0618 |
Location: |
Chr.5:135376140-135404733 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pom121 | NM_148932.2 | NP_683734.2 | ||
| AF516680.1 | AAM64199.1 | |||
| AK154473.1 | BAE32610.1 | |||
| AK172982.1 | BAD32260.1 | |||
| BC053101.1 | AAH53101.1 |
More Information
Set Membership: |
|

Genomic Map