Assay Details
Target Gene Details
Entrez Gene ID: | 54153 |
Gene Name: | RAS p21 protein activator 4 |
Gene Aliases: |
AA793972, AW112107, BB079060, CAPRI, GAPL |
Location: |
Chr.5:136083916-136111860 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rasa4 | NM_001039103.3 | 15 | 1795 | NP_001034192.1 |
| NM_133914.3 | 16 | 1933 | NP_598675.2 | |
| AK014220.1 | 15 | 2082 | ||
| AK044762.1 | 14 | 1878 | BAC32072.1 | |
| AK133565.1 | 15 | 2082 | ||
| AK154335.1 | 16 | 1934 | BAE32521.1 | |
| AK155146.1 | 16 | 1934 | BAE33076.1 | |
| AK220218.1 | 16 | 1809 | BAD90143.1 | |
| AY591339.1 | 16 | 1834 | AAT00515.1 | |
| BC057460.1 | 16 | 1787 | AAH57460.1 | |
| DQ317932.1 | 15 | 1679 | ABC47038.1 |
More Information
Set Membership: |
|

Genomic Map