Assay Details
Target Gene Details
Entrez Gene ID: | 54153 |
Gene Name: | RAS p21 protein activator 4 |
Gene Aliases: |
AA793972, AW112107, BB079060, CAPRI, GAPL |
Location: |
Chr.5:136083916-136111860 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rasa4 | NM_001039103.3 | NP_001034192.1 | ||
| NM_133914.3 | NP_598675.2 | |||
| AK014220.1 | ||||
| AK044762.1 | BAC32072.1 | |||
| AK133565.1 | ||||
| AK154335.1 | BAE32521.1 | |||
| AK155146.1 | BAE33076.1 | |||
| AK220218.1 | BAD90143.1 | |||
| AY591339.1 | AAT00515.1 | |||
| BC057460.1 | AAH57460.1 | |||
| BY195753.1 | ||||
| DQ317932.1 | ABC47038.1 |
More Information
Set Membership: |
|

Genomic Map