Assay Details
Target Gene Details
Entrez Gene ID: | 19699 |
Gene Name: | reelin |
Gene Aliases: |
reeler, rl |
Location: |
Chr.5:21884454-22344705 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 58 - Exon 58 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Reln | NM_011261.2 | NP_035391.2 | ||
| AK017094.1 | BAB30592.1 | |||
| AK047608.1 | ||||
| BC118041.1 | ||||
| D63520.1 | BAA09788.1 | |||
| U24703.1 | AAB91599.1 |
More Information
Set Membership: |
|

Genomic Map