Assay Details
Target Gene Details
Entrez Gene ID: | 243197 |
Gene Name: | major facilitator superfamily domain containing 7A |
Gene Aliases: |
4732482E20Rik, Mfsd7 |
Location: |
Chr.5:108441052-108450260 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mfsd7a | NM_172883.2 | 3 | 482 | NP_766471.1 |
| AK029021.1 | 3 | 482 | BAC26248.1 | |
| AK087226.1 | 3 | 692 | ||
| AK171069.1 | 2 | 3114 | ||
| BC094512.1 | AAH94512.1 |
More Information
Set Membership: |
|

Genomic Map