Assay Details
Target Gene Details
Entrez Gene ID: | 17766 |
Gene Name: | nudix (nucleoside diphosphate linked moiety X)-type motif 1 |
Gene Aliases: |
Mth1 |
Location: |
Chr.5:140321627-140338137 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nudt1 | NM_008637.1 | NP_032663.1 | ||
| AK011695.1 | BAB27785.1 | |||
| AK088309.1 | BAC40274.1 | |||
| AK168312.1 | BAE40252.1 | |||
| BC021940.1 | AAH21940.1 | |||
| BC098239.1 | AAH98239.1 | |||
| D49956.1 | BAA08711.1 |
More Information
Set Membership: |
|

Genomic Map