Assay Details
Target Gene Details
Entrez Gene ID: | 231602 |
Gene Name: | purinergic receptor P2X, ligand-gated ion channel, 2 |
Gene Aliases: |
P2X2a, P2x2 |
Location: |
Chr.5:110339812-110343446 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| P2rx2 | NM_001164833.1 | NP_001158305.1 | ||
| NM_001164834.1 | NP_001158306.1 | |||
| NM_153400.4 | NP_700449.2 | |||
| AB094663.1 | BAC55013.1 | |||
| AB094664.1 | BAC55014.1 | |||
| AK141196.1 | BAE24579.1 | |||
| AK158334.1 | BAE34462.1 | |||
| AY044240.2 | AAK95327.2 | |||
| BC106963.1 | AAI06964.1 | |||
| BC106964.1 | AAI06965.1 |
More Information
Set Membership: |
|

Genomic Map