Assay Details
Target Gene Details
Entrez Gene ID: | 18438 |
Gene Name: | purinergic receptor P2X, ligand-gated ion channel 4 |
Gene Aliases: |
AI504491, AW555605, D5Ertd444e, P2X4 |
Location: |
Chr.5:122707504-122729739 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| P2rx4 | NM_011026.2 | 1 | 201 | NP_035156.2 |
| AF089751.1 | 1 | 135 | AAC95601.1 | |
| AF089752.1 | 1 | 135 | AAC95602.1 | |
| AJ251459.1 | 1 | 115 | CAB90749.1 | |
| AJ251460.1 | 1 | 115 | CAB90750.1 | |
| AJ251461.1 | 1 | 115 | CAB90751.1 | |
| AJ251462.1 | 1 | 115 | CAB90752.1 | |
| AK041904.1 | 1 | 189 | ||
| AK150351.1 | 1 | 191 | BAE29488.1 | |
| AK153427.1 | 1 | 142 | ||
| AK162498.1 | 1 | 186 | ||
| AK162820.1 | 1 | 167 | ||
| AK163104.1 | 1 | 189 | BAE37194.1 | |
| BC005597.1 | 1 | 149 | AAH05597.1 | |
| BY150918.1 | 1 | 215 | ||
| JX417451.1 | 1 | 161 | ||
| JX674050.1 |
More Information
Set Membership: |
|

Genomic Map