Assay Details
Target Gene Details
Entrez Gene ID: | 320951 |
Gene Name: | phosphatidylserine decarboxylase |
Gene Aliases: |
9030221M09Rik |
Location: |
Chr.5:32736303-32785636 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pisd | NM_177298.3 | 3 | 285 | NP_796272.2 |
| AK033483.1 | 3 | 277 | BAC28313.1 | |
| AK034656.1 | 3 | 287 | BAC28786.1 | |
| AK162028.1 | 3 | 278 | BAE36689.1 | |
| AK162737.1 | 3 | 286 | BAE37044.1 | |
| AK167554.1 | 2 | 227 | BAE39619.1 | |
| BC070408.1 | 3 | 284 | AAH70408.1 | |
| BC094594.1 | 3 | 276 | AAH94594.1 | |
| BC116695.1 | 3 | 287 | ||
| BC117509.1 | 3 | 287 | ||
| CX201846.1 | 3 | 300 |
More Information
Set Membership: |
|

Genomic Map