Assay Details
Target Gene Details
Entrez Gene ID: | 73078 |
Gene Name: | peptidase (mitochondrial processing) beta |
Gene Aliases: |
3110004O18Rik, MPP11, MPPB, MPPP52 |
Location: |
Chr.5:21737160-21757152 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pmpcb | NM_028431.2 | NP_082707.1 | ||
| AK013995.1 | BAB29105.1 | |||
| AK152673.1 | ||||
| AK169417.1 | BAE41163.1 |
More Information
Set Membership: |
|

Genomic Map