Assay Details
Target Gene Details
Entrez Gene ID: | 212996 |
Gene Name: | Williams-Beuren syndrome chromosome region 17 homolog (human) |
Gene Aliases: |
AA388321, E330012B09Rik |
Location: |
Chr.5:130874951-131307522 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Wbscr17 | NM_145218.3 | NP_660253.2 | ||
| AB175684.1 | BAD91810.1 | |||
| AF467979.1 | AAM62404.1 | |||
| BC158110.1 |
More Information
Set Membership: |
|

Genomic Map