Assay Details
Target Gene Details
Entrez Gene ID: | 212996 |
Gene Name: | Williams-Beuren syndrome chromosome region 17 homolog (human) |
Gene Aliases: |
AA388321, E330012B09Rik |
Location: |
Chr.5:130874951-131307522 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Wbscr17 | NM_145218.3 | 6 | 1947 | NP_660253.2 |
| AB175684.1 | 6 | 1025 | BAD91810.1 | |
| AF467979.1 | 6 | 1228 | AAM62404.1 | |
| AK035817.1 | 6 | 1701 | BAC29197.1 | |
| AK048758.1 | 6 | 1692 | BAC33446.2 | |
| AK051281.1 | 6 | 2347 | BAC34591.2 | |
| BC158110.1 | 6 | 1974 |
More Information
Set Membership: |
|

Genomic Map